Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Primary Calculator
SCI_ED

Catalog No. s79008
Click to view available options
Display Type:
Digital
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.

Includes

Three-key memory, auto shutoff, eight-digit display, one AA battery (S43921H)

Learning calculator skills is easy.

  • Bright blue calculator is both solar and battery powered
  • Students start with basic math operations and work their way up to more advanced concepts such as percents and square roots
  • Perfect for independent math practice
  • Dimensions (W × H): 2.5 × 4.5 in. (6.25 × 11.3cm)

Specifications

Product Type Primary Calculator
Display Type Digital
Color Bright Blue
Display Dimensions 2.5 x 4.5 in.
Memory 3-key
Quantity 10/Pk.
Recommended Age 3 to 17
Height (English) 4.5 in.
Height (Metric) 11.4cm
Width (English) 2.5 in.
Width (Metric) 6.3cm
Power Supply Battery, Solar
For Use With (Application) Perfect for independent math practice
Includes Three-key memory, auto shutoff, eight-digit display, one AA battery (S43921H)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.