Learn More
Promocell GmbH PromoCell Primary Cancer Culture System contains: C-28080, C-43080 250 ml, with C-39880 attached to flask
Catalog No. NC1709727
Encompass
PromoCell’s Primary Cancer Culture System is a standardized, defined and animal component-free (D-ACF) system for the isolation and culture of human primary tumor cells. The medium is a part of the cancer media toolbox, a complete cell culture solution for the isolation and long-term culture of cancer cells. The PCCS is ideal for CSCs from human primary tumor biopsies, patient-derived xenografts (PDX), or commercially available cancer cell lines.Designed to specifically isolate and maintain CSC, the PCCS supports the aberrant metabolic traits of malignant cells. As this principle is not tissue-specific, the PCCS can be used to isolate CSCs from most tumor types.Key features:Perform up to five CSC isolations with one medium kitGain access to pure and stable CSC cultures from primary tumors, patient-derived xenografts (PDX), and established cell linesUniversal support for most tumor typesEnables long-term growth support and multiple passaging
Catalog No. NC1709727 Supplier Promocell GmbH Supplier No. C28081
PromoCell’s Primary Cancer Culture System is a standardized, defined and animal component-free (D-ACF) system for the isolation and culture of human primary tumor cells. The medium is a part of the cancer media toolbox, a complete cell culture solution for the isolation and long-term culture of cancer cells. The PCCS is ideal for CSCs from human primary tumor biopsies, patient-derived xenografts (PDX), or commercially available cancer cell lines.Designed to specifically isolate and maintain CSC, the PCCS supports the aberrant metabolic traits of malignant cells. As this principle is not tissue-specific, the PCCS can be used to isolate CSCs from most tumor types.Key features:Perform up to five CSC isolations with one medium kitGain access to pure and stable CSC cultures from primary tumors, patient-derived xenografts (PDX), and established cell linesUniversal support for most tumor typesEnables long-term growth support and multiple passaging