Learn More
Lonza Walkersville Primary Cell Optimization 96-well Nucleofector Kit
Catalog No. 501131865
Encompass
The Primary Cell Optimization Nucleofector Kit is the ideal tool to conveniently and rapidly determine Nucleofection conditions for primary cell types lacking an Optimized Protocol for the 4D-Nucleofector 96-well Unit.Applications:Determination of Nucleofection conditions for primary cell types lacking an Optimized Protocol for the 4D-Nucleofector 96-well UnitContent:0.675 mL Nucleofector Solution P10.675 mL Nucleofector Solution P20.675 mL Nucleofector Solution P30.675 mL Nucleofector Solution P40.675 mL Nucleofector Solution P54 x 0.15 mL Supplement 11 x 0.15 mL Supplement 32 x 50 µg pmaxGFP Vector (1 μg/μl in 10 mM Tris pH 8.0) 2 x 96-well Nucleocuvette Plates
Catalog No. 50-113-1865 Supplier Lonza Walkersville Supplier No. V4SP9096
The Primary Cell Optimization Nucleofector Kit is the ideal tool to conveniently and rapidly determine Nucleofection conditions for primary cell types lacking an Optimized Protocol for the 4D-Nucleofector 96-well Unit.Applications:Determination of Nucleofection conditions for primary cell types lacking an Optimized Protocol for the 4D-Nucleofector 96-well UnitContent:0.675 mL Nucleofector Solution P10.675 mL Nucleofector Solution P20.675 mL Nucleofector Solution P30.675 mL Nucleofector Solution P40.675 mL Nucleofector Solution P54 x 0.15 mL Supplement 11 x 0.15 mL Supplement 32 x 50 µg pmaxGFP Vector (1 μg/μl in 10 mM Tris pH 8.0) 2 x 96-well Nucleocuvette Plates