Learn More
Lonza Walkersville Primary Cell Optimization 4D-Nucleofector X Kit
Catalog No. NC0836140
Encompass
The Primary Cell Optimization 4D-Nucleofector X Kit is the ideal tool to conveniently and rapidly determine Nucleofection conditions for primary cell types lacking an Optimized Protocol for the 4D-Nucleofector X Unit.Applications:Determination of Nucleofection conditions for primary cell types lacking an Optimized Protocol for the 4D-Nucleofector X UnitContent:0.675 mL Nucleofector Solution P10.675 mL Nucleofector Solution P20.675 mL Nucleofector Solution P30.675 mL Nucleofector Solution P40.675 mL Nucleofector Solution P54 x 0.15 mL Supplement 11 x 0.15 mL Supplement 350 µg pmaxGFP Vector (1 μg/μl in 10 mM Tris pH 8.0)6 x 20 µL 16-well Nucleocuvette Strips
Catalog No. NC0836140 Supplier Lonza Walkersville Supplier No. V4XP9096
The Primary Cell Optimization 4D-Nucleofector X Kit is the ideal tool to conveniently and rapidly determine Nucleofection conditions for primary cell types lacking an Optimized Protocol for the 4D-Nucleofector X Unit.Applications:Determination of Nucleofection conditions for primary cell types lacking an Optimized Protocol for the 4D-Nucleofector X UnitContent:0.675 mL Nucleofector Solution P10.675 mL Nucleofector Solution P20.675 mL Nucleofector Solution P30.675 mL Nucleofector Solution P40.675 mL Nucleofector Solution P54 x 0.15 mL Supplement 11 x 0.15 mL Supplement 350 µg pmaxGFP Vector (1 μg/μl in 10 mM Tris pH 8.0)6 x 20 µL 16-well Nucleocuvette Strips