Learn More
Waters Corp Waters Primary Check Valve, Stainless Steel, 1/pk
Catalog No. 502224462 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement primary check valve is a spring loaded component used in ACQUITY Arc and Arc HPLC Quaternary Solvent Managers, and can also be used with NanoACQUITY Binary, UPC2 Binary, and ACQUITY Isocratic Solvent Managers. It ensures reliable operation and performance maintenance for various chromatographic systems.
  • Component: ACQUITY UPLC ISM, ACQUITY UPLC BSM, ACQUITY UPLC QSM, ACQUITY UPLC ASM, nanoACQUITY ASM, nanoACQUITY BSM
  • Product type: Rebuild kits, performance maintenance kits
  • Units per package: 1 pk
  • Type: Check valve, pump head, performance maintenance, rebuild kit
  • System type: UPLC
  • Instrument: ACQUITY Arc HPLC system, ACQUITY UPLC H-Class system, nanoACQUITY UPLC system
  • Material: Stainless steel
  • Replacement schedule: Annual
Catalog No. 50-222-4462 Supplier Waters Corp Supplier No. 700010664
This replacement primary check valve is a spring loaded component used in ACQUITY Arc and Arc HPLC Quaternary Solvent Managers, and can also be used with NanoACQUITY Binary, UPC2 Binary, and ACQUITY Isocratic Solvent Managers. It ensures reliable operation and performance maintenance for various chromatographic systems.
  • Component: ACQUITY UPLC ISM, ACQUITY UPLC BSM, ACQUITY UPLC QSM, ACQUITY UPLC ASM, nanoACQUITY ASM, nanoACQUITY BSM
  • Product type: Rebuild kits, performance maintenance kits
  • Units per package: 1 pk
  • Type: Check valve, pump head, performance maintenance, rebuild kit
  • System type: UPLC
  • Instrument: ACQUITY Arc HPLC system, ACQUITY UPLC H-Class system, nanoACQUITY UPLC system
  • Material: Stainless steel
  • Replacement schedule: Annual