Learn More
ATCC Primary Corneal Epithelial Cells; Normal Human
Catalog No. 502383519
Encompass
ATCC's Normal Human Primary Corneal Epithelial Cells provide an ideal cell system to propagate corneal epithelial cells in serum-free conditions when grown in Corneal Epithelial Cell Basal Media supplemented with Corneal Epithelial Cell Growth Kit components. The cells are cryopreserved at the second passage to ensure the highest viability and plating efficiency. ATCC Primary Cell Solutions cells, media, supplements, and reagents are quality tested together to guarantee optimum performance and reliability.
Catalog No. 50-238-3519 Supplier ATCC Supplier No. PCS700010
 Restricted Item
Add to Cart
Add to Cart
ATCC's Normal Human Primary Corneal Epithelial Cells provide an ideal cell system to propagate corneal epithelial cells in serum-free conditions when grown in Corneal Epithelial Cell Basal Media supplemented with Corneal Epithelial Cell Growth Kit components. The cells are cryopreserved at the second passage to ensure the highest viability and plating efficiency. ATCC Primary Cell Solutions cells, media, supplements, and reagents are quality tested together to guarantee optimum performance and reliability.