Learn More
ATCC Primary Coronary Artery Endothelial Cells; Normal, Human
Supplier:  ATCC PCS100020
Encompass
ATCC Normal Human Primary Coronary Artery Endothelial Cells, when grown in Vascular Cell Basal Media supplemented with Endothelial Cell Growth Kit components, provide an ideal cell system to propagate coronary artery endothelial cells in low serum conditions with or without the addition of human recombinant VEGF.
Catalog No. 50-238-4449