Learn More
Motic™ Swift Line Primary Education Compound Microscope
SCI_ED
Catalog No. S81373 Shop All Fisherbrand Products
Change view
Click to view available options
Magnification Power:
40X, 100X, 400X
Catalog No. Magnification Power
S81373 40X, 100X, 400X
1 options
Catalog No. S81373
Add to Cart
Add to Cart
Motic Swift Line Primary Education Compound Microscope packs full-sized performance in a compact microsocpe scaled for younger students.
  • Widefield 10X eyepiece with revolving pointer, 45° inclined for viewing ease when seated
  • Monocular head rotates 360° for easy repositioning of eyepiece and sharing by more than one student
  • Achromatic, color-coded 4X, 10X, 40XR objectives
  • Oversized focus knob provides mid-range focusing between normal coarse and fine focus
  • Locked-on spring activated stage clips
  • Substage disc diaphragm
Specifications
Eyepiece WF 10X
Head Style Monocular
Lighting Type LED
Magnification Power 40X, 100X, 400X
Objective Types Achromatic
Product Type Beginner Microscope
For Use With (Application) Primary Science Education
Voltage 110/220 V
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov