Learn More
ATCC Primary Epidermal Melanocytes; Normal, Human, Neonatal
Supplier:  ATCC PCS200012
Encompass
Primary Epidermal Melanocytes; Normal, Human, Neonatal (HEMn) is a melanocyte isolated from the skin of a donor and has applications in melanoma, response to UV radiation, psoriasis, and other skin diseases, skin trauma (eg, wound repair, scars, burns), cosmetic research (eg, skin lightening compounds, skin protecting compounds).
Catalog No. 50-238-4148