Learn More
ATCC Primary Epidermal Melanocytes; Normal, Human, Adult
Supplier:  ATCC PCS200013
Encompass
Normal Human Primary Epidermal Melanocytes from Adult donors is a melanocyte isolated from the skin of a donor and has applications in melanoma, response to UV radiation, psoriasis, and other skin diseases, skin trauma (eg, wound repair, scars, burns), cosmetic research (eg, skin lightening compounds, skin protecting compounds).
Catalog No. 50-238-4543