Learn More
STEMCELL Technologies Human Peripheral Blood CD4+CD25+CD127low T cells, Frozen, Size: 1 x 10^6 cells
SDP
Catalog No. 502045572
Encompass
Primary human CD4+CD25+CD127low T cells were isolated from peripheral blood using immunomagnetic separation techniques. CD4+, CD25+, and CD127low are expressed on specific subsets of T cells.Cells were obtained using Institutional Review Board (IRB)-approved consent forms and protocols.
Catalog No. 50-204-5572 Supplier STEMCELL Technologies Supplier No. 2000122
Add to Cart
Add to Cart
Primary human CD4+CD25+CD127low T cells were isolated from peripheral blood using immunomagnetic separation techniques. CD4+, CD25+, and CD127low are expressed on specific subsets of T cells.Cells were obtained using Institutional Review Board (IRB)-approved consent forms and protocols.