Learn More
STEMCELL Technologies Human Peripheral Blood CD4+CD25- T cells (Responders), Frozen, Size: 2 x 10^6 cells
SDP
Catalog No. 502045575
Encompass
Primary human CD4+CD25- T cells were isolated from peripheral blood using immunomagnetic separation techniques. CD4+CD25- T cells are often refered to as responder T cells.Cells were obtained using Institutional Review Board (IRB)-approved consent forms and protocols.
Catalog No. 50-204-5575 Supplier STEMCELL Technologies Supplier No. 2000125
Primary human CD4+CD25- T cells were isolated from peripheral blood using immunomagnetic separation techniques. CD4+CD25- T cells are often refered to as responder T cells.Cells were obtained using Institutional Review Board (IRB)-approved consent forms and protocols.