Learn More
STEMCELL Technologies Human Peripheral Blood CD4+CD25+CD127lowFOXP3+ T cells (Tregs), Frozen , Size: 1 x 10^6 cells
SDP
Catalog No. 502045570
Encompass
Primary human CD4+CD25+CD127lowFOXP3+ T cells were isolated from peripheral blood using immunomagnetic separation techniques. These cells are often referred to as regulatory T cells and are used in immune response assays. Cells were obtained using Institutional Review Board (IRB)-approved consent forms and protocols.
Catalog No. 50-204-5570 Supplier STEMCELL Technologies Supplier No. 2000120
Add to Cart
Add to Cart
Request a Quote
Primary human CD4+CD25+CD127lowFOXP3+ T cells were isolated from peripheral blood using immunomagnetic separation techniques. These cells are often referred to as regulatory T cells and are used in immune response assays. Cells were obtained using Institutional Review Board (IRB)-approved consent forms and protocols.