Learn More
STEMCELL Technologies Human Peripheral Blood Plasma, Fresh, Size: 50 mL
SDP
Catalog No. 502069402
Encompass
Primary human plasma has been isolated from peripheral blood (PB) using centrifugation. PB was collected using acid-citrate-dextrose solution A (ACDA). Human plasma is the liquid component of blood containing water, salts, lipids, hormones, proteins (including albumin), immunoglobulins, clotting factors, and fibrinogen. It does not contain the cellular components of blood—the erythrocytes and the buffy coat composed of leukocytes and platelets. Plasma, from a single donor, was obtained using Institutional Review Board (IRB)-approved consent forms and protocols.Certain products are only available in select territories. Please contact your local Sales representative or Product & Scientific Support at techsupport@stemcell.com for further information.Browse our frequently asked questions (FAQs) on primary cells. (https://www.stemcell.com/technical-resources/educational-materials/tech-tips/primary-cells-faq.html)
Catalog No. 50-206-9402 Supplier STEMCELL Technologies Supplier No. 2000154
Primary human plasma has been isolated from peripheral blood (PB) using centrifugation. PB was collected using acid-citrate-dextrose solution A (ACDA). Human plasma is the liquid component of blood containing water, salts, lipids, hormones, proteins (including albumin), immunoglobulins, clotting factors, and fibrinogen. It does not contain the cellular components of blood—the erythrocytes and the buffy coat composed of leukocytes and platelets. Plasma, from a single donor, was obtained using Institutional Review Board (IRB)-approved consent forms and protocols.Certain products are only available in select territories. Please contact your local Sales representative or Product & Scientific Support at techsupport@stemcell.com for further information.Browse our frequently asked questions (FAQs) on primary cells. (https://www.stemcell.com/technical-resources/educational-materials/tech-tips/primary-cells-faq.html)