Learn More
STEMCELL Technologies Human Peripheral Blood Monocytes, Frozen, Size: 2 x 10^7 cells
SDP
Catalog No. 502069413
Encompass
Peripheral blood (PB) primary human monocytes are isolated from mononuclear cells (MNCs) using the negative selection immunomagnetic technique. Monocytes are untouched and ready for any downstream application. PB was collected using the anticoagulant acid-citrate-dextrose solution A (ACDA).Cells were obtained using Institutional Review Board (IRB)-approved consent forms and protocols.Certain products are only available in select territories. Please contact your local Sales representative or Product & Scientific Support at techsupport@stemcell.com for further information.Browse our Frequently Asked Questions (FAQs) on Primary Cells.
Catalog No. 50-206-9413 Supplier STEMCELL Technologies Supplier No. 2000166
Peripheral blood (PB) primary human monocytes are isolated from mononuclear cells (MNCs) using the negative selection immunomagnetic technique. Monocytes are untouched and ready for any downstream application. PB was collected using the anticoagulant acid-citrate-dextrose solution A (ACDA).Cells were obtained using Institutional Review Board (IRB)-approved consent forms and protocols.Certain products are only available in select territories. Please contact your local Sales representative or Product & Scientific Support at techsupport@stemcell.com for further information.Browse our Frequently Asked Questions (FAQs) on Primary Cells.