Learn More
McKesson General Medical PRIMARY IV ADMINISTRATION SET
Supplier:  McKesson General Medical 467609
Encompass
Primary IV Administration Set Clearlink™ 10 Drops / mL Drip Rate 106 Inch Tubing 1 Port
Catalog No. NC2241312