Learn More
ATCC Primary Lung Fibroblast Cells;Fibrosis,Human
Catalog No. 502382289
Encompass
Primary lung fibroblast are spindle-shaped cells isolated from the respiratory tract of a donor with fibrosis. The cells have an application in microbial (eg, viral, bacterial) infection and pathogenesis, airway inflammation and wound healing, asthma, pulmonary fibrosis, chronic obstructive pulmonary disease, chronic bronchitis, toxicology, and testing of pharmaceuticals.
Catalog No. 50-238-2289 Supplier ATCC Supplier No. PCS201020
Primary lung fibroblast are spindle-shaped cells isolated from the respiratory tract of a donor with fibrosis. The cells have an application in microbial (eg, viral, bacterial) infection and pathogenesis, airway inflammation and wound healing, asthma, pulmonary fibrosis, chronic obstructive pulmonary disease, chronic bronchitis, toxicology, and testing of pharmaceuticals.