Learn More
ATCC Primary Lung Fibroblasts; Asthma, Human
Catalog No. 502384539
Encompass
Primary lung fibroblasts are spindle-shaped cells isolated from the respiratory tract of a donor with asthma. The cells have an application in microbial (eg, viral, bacterial) infection and pathogenesis, airway inflammation and wound healing, asthma, pulmonary fibrosis, chronic obstructive pulmonary disease, chronic bronchitis, toxicology, and testing of pharmaceuticals.
Catalog No. 50-238-4539 Supplier ATCC Supplier No. PCS201015
Primary lung fibroblasts are spindle-shaped cells isolated from the respiratory tract of a donor with asthma. The cells have an application in microbial (eg, viral, bacterial) infection and pathogenesis, airway inflammation and wound healing, asthma, pulmonary fibrosis, chronic obstructive pulmonary disease, chronic bronchitis, toxicology, and testing of pharmaceuticals.