Learn More
ATCC Primary Lung Fibroblasts; COPD, Human
Catalog No. 502384544
Encompass
Primary lung fibroblasts COPD are spindle-shaped cells isolated from the respiratory tract of a donor with COPD. The cells have an application in microbial (eg, viral, bacterial) infection and pathogenesis, airway inflammation and wound healing, asthma, pulmonary fibrosis, chronic obstructive pulmonary disease, chronic bronchitis, toxicology, and testing of pharmaceuticals.
Catalog No. 50-238-4544 Supplier ATCC Supplier No. PCS201017
Primary lung fibroblasts COPD are spindle-shaped cells isolated from the respiratory tract of a donor with COPD. The cells have an application in microbial (eg, viral, bacterial) infection and pathogenesis, airway inflammation and wound healing, asthma, pulmonary fibrosis, chronic obstructive pulmonary disease, chronic bronchitis, toxicology, and testing of pharmaceuticals.