Learn More
ATCC Primary Peripheral Blood CD14+ Monocytes, Normal, Human
Catalog No. 502382004
Encompass
Primary Peripheral Blood CD14+ Monocytes are primary peripheral blood mononuclear cells isolated from normal blood tissue that have been fully authenticated and characterized by ATCC. These cells can be used in the study of immunology, infection, cancer, hematology, and T-cell suppression assay. One vial contains a minimum of 5.0 × 105 viable cells (provided).
Catalog No. 50-238-2004 Supplier ATCC Supplier No. PCS800010
Primary Peripheral Blood CD14+ Monocytes are primary peripheral blood mononuclear cells isolated from normal blood tissue that have been fully authenticated and characterized by ATCC. These cells can be used in the study of immunology, infection, cancer, hematology, and T-cell suppression assay. One vial contains a minimum of 5.0 × 105 viable cells (provided).