Learn More
ATCC Primary Peripheral Blood: CD8+ Cytotoxic T Cells; Normal; Human
Catalog No. 502382864
Encompass
Primary CD8+ Cytotoxic T Cells are lymphocytes that regulate and/or carry out the body’s immune response by eliminating intracellular pathogens, such as viruses and some bacteria and parasites. This cell line was isolated from normal blood and can be used in research, compound screening, and immune function assays.
Catalog No. 50-238-2864 Supplier ATCC Supplier No. PCS800017
Primary CD8+ Cytotoxic T Cells are lymphocytes that regulate and/or carry out the body’s immune response by eliminating intracellular pathogens, such as viruses and some bacteria and parasites. This cell line was isolated from normal blood and can be used in research, compound screening, and immune function assays.