Learn More
ATCC Primary Peripheral Blood Mononuclear Cells, Normal, Human
Catalog No. 502383396
Encompass
Primary Peripheral Blood Mononuclear Cells (PBMC) are a heterogeneous population of blood cells with a single round nucleus and include macrophages, dendritic cells, monocytes and lymphocytes. These cells were isolated from normal blood tissue and have been fully authenticated and characterized by ATCC. The cells can be used in the study of immunology, infection, cancer, hematology, and t-cell suppression assay. This one vial contains a minimum of 25 × 106 viable cells (provided).
Catalog No. 50-238-3396 Supplier ATCC Supplier No. PCS800011
Primary Peripheral Blood Mononuclear Cells (PBMC) are a heterogeneous population of blood cells with a single round nucleus and include macrophages, dendritic cells, monocytes and lymphocytes. These cells were isolated from normal blood tissue and have been fully authenticated and characterized by ATCC. The cells can be used in the study of immunology, infection, cancer, hematology, and t-cell suppression assay. This one vial contains a minimum of 25 × 106 viable cells (provided).