Learn More
ATCC Primary Pulmonary Artery; Smooth Muscle Cells; Normal, Human
Supplier:  ATCC PCS100023
Encompass
Normal primary pulmonary artery smooth muscle cells are elongated and sometimes triangular in shape and isolated from a pulmonary artery of a donor. The cells can be used in hypertension, arterial disease, organ-specific drug screening, and vascular differentiation.
Catalog No. 50-238-4523