Learn More
ATCC Primary Renal Mixed Epithelial Cells; Normal, Human
Catalog No. 502384665
Encompass
ATCC Normal Human Primary Renal Mixed Epithelial Cells provide an ideal cell system to propagate renal epithelial cells in low serum (0.5% FBS) conditions when grown in Renal Epithelial Cell Basal Media supplemented with Renal Epithelial Cell Growth Kit components. The cells are cryopreserved in the first passage to ensure the highest viability and plating efficiency. ATCC Primary Cell Solutions cells, media, supplements and reagents are quality tested together to guarantee optimum performance and reliability.
Catalog No. 50-238-4665 Supplier ATCC Supplier No. PCS400012
ATCC Normal Human Primary Renal Mixed Epithelial Cells provide an ideal cell system to propagate renal epithelial cells in low serum (0.5% FBS) conditions when grown in Renal Epithelial Cell Basal Media supplemented with Renal Epithelial Cell Growth Kit components. The cells are cryopreserved in the first passage to ensure the highest viability and plating efficiency. ATCC Primary Cell Solutions cells, media, supplements and reagents are quality tested together to guarantee optimum performance and reliability.