Learn More
Description
- Experiment and explore with real science tools, perfectly sized for little hands.
- This colorful set encourages early hands-on science investigations at the kitchen table.
- 22-pieces include: beaker, magnifying glass, funnel, test tubes and more, all made of durable plastic.
- Also includes activity guide and cards with fun, easy-to-do experiments.
Specifications
Specifications
| Grade Level | Elementary, Intermediate - Middle School, High School |
| Grade | K to 12 |
| Class Size | 1 Student |
| Product Type | STEAM Learning |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
