Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Learning Resources Primary Science Magnetic Wands, Set of 24
SCI_ED

Catalog No. S02763
Change view
Click to view available options
Class Size:
24 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S02763 24 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02763 Supplier Learning Resources Supplier No. LER0805
Add to Cart
Add to Cart

Talk about attraction! Wands in 6 colors engage students in hands-on magnetism activities. Great for physical science centers. Helps children learn about magnetic properties, sharpen observation skills, and more.

  • Wands in 6 colors engage students in hands-on magnetism activities.
  • Great for physical science centers.
  • Helps children learn about magnetic properties, sharpen observation skills, and more.
  • Learning Resources Magnetic Wands, Set of 24
  • Features: 24 plastic wands that measure 8 in. L each Plastic storage bucket with lid

Specifications

Grade Level Elementary, Intermediate - Middle School, High School
Grade K to 12
Class Size 24 Students
Product Type Magnetic Wands
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.