Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Primary Science Sensory Tubes
SCI_ED

Catalog No. s04656
Change view
Click to view available options
Grade:
K
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S04656 K
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.
Catalog No. S04656 Supplier Learning Resources, Inc Supplier No. LER2445
Only null left
Add to Cart
Add to Cart
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.

Clear tubes nurture sensory development and science exploration.

  • Set of four tubes
  • Two easy-twist, solid lids hold liquid securely inside
  • Four vented lids allow smell or live creature exploration
  • Measure 12 × 2.5 in.
  • Grades K+

Specifications

Grade Level Elementary
Grade K
Age 2 to 12
Height (English) 12 in.
For Use With (Application) Nurture sensory development and science exploration
Diameter (English) Outer 2.5 in.
Product Type Primary Science Sensory Tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.