Learn More
ATCC Primary Skeletal Muscle Cell Growth Kit
Catalog No. 502381901
Encompass
Primary Skeletal Muscle Cell Growth Kit contains components that when added to Mesenchymal Stem Cell Basal Medium (ATCC PCS-500-030) create a complete ATCC Primary Cell Solutions culture environment for skeletal muscle cells derived from a normal human skeletal tissue (eg, Primary Skeletal Muscle Cells, Normal, Human, ATCC PCS-950-010). The medium formulation is designed to support normal skeletal cell proliferation and growth. No feeder layers, extracellular matrix proteins, or other substrates are required.
Catalog No. 50-238-1901 Supplier ATCC Supplier No. PCS950040
 Restricted Item
Add to Cart
Add to Cart
Primary Skeletal Muscle Cell Growth Kit contains components that when added to Mesenchymal Stem Cell Basal Medium (ATCC PCS-500-030) create a complete ATCC Primary Cell Solutions culture environment for skeletal muscle cells derived from a normal human skeletal tissue (eg, Primary Skeletal Muscle Cells, Normal, Human, ATCC PCS-950-010). The medium formulation is designed to support normal skeletal cell proliferation and growth. No feeder layers, extracellular matrix proteins, or other substrates are required.