Learn More
ATCC Primary Small Airway Epithelial Cells; COPD, Human
Catalog No. 502381853
Encompass
Primary small airway epithelial cells are spindle-shaped cells isolated from the respiratory tract of a donor with COPD. The cells have an application in microbial (eg, viral, bacterial) infection and pathogenesis, airway inflammation and wound healing, asthma, pulmonary fibrosis, chronic obstructive pulmonary disease, chronic bronchitis, toxicology, and testing of pharmaceuticals.
Catalog No. 50-238-1853 Supplier ATCC Supplier No. PCS301013
Primary small airway epithelial cells are spindle-shaped cells isolated from the respiratory tract of a donor with COPD. The cells have an application in microbial (eg, viral, bacterial) infection and pathogenesis, airway inflammation and wound healing, asthma, pulmonary fibrosis, chronic obstructive pulmonary disease, chronic bronchitis, toxicology, and testing of pharmaceuticals.