Learn More
ATCC Primary Small Airway Epithelial Cells; Asthma, Human
Catalog No. 502382579
Encompass
Primary small airway epithelial cells are spindle-shaped that were isolated from the respiratory tract of a donor with asthma. The cells have an application in microbial (eg, viral, bacterial) infection and pathogenesis, airway inflammation and wound healing, asthma, pulmonary fibrosis, chronic obstructive pulmonary disease, chronic bronchitis, toxicology, and testing of pharmaceuticals.
Catalog No. 50-238-2579 Supplier ATCC Supplier No. PCS301011
Primary small airway epithelial cells are spindle-shaped that were isolated from the respiratory tract of a donor with asthma. The cells have an application in microbial (eg, viral, bacterial) infection and pathogenesis, airway inflammation and wound healing, asthma, pulmonary fibrosis, chronic obstructive pulmonary disease, chronic bronchitis, toxicology, and testing of pharmaceuticals.