Learn More
ATCC Primary Small Airway Epithelial; Cells, Fibrosis;Human (Homo sapiens)
Supplier:  ATCC PCS301014
Encompass
Primary small airway epithelial cells were isolated from the respiratory tract of a donor with fibrosis. The cells have an application in microbial (eg, viral, bacterial) infection and pathogenesis, airway inflammation and wound healing, asthma, pulmonary fibrosis, chronic obstructive pulmonary disease, chronic bronchitis, toxicology, and testing of pharmaceuticals.
Catalog No. 50-238-4226