Learn More
ATCC Primary Small Airway Epithelial Cells; Normal, Human
Catalog No. 502384580
Encompass
Primary Small Airway Epithelial Cells; Normal, Human (HSAEC) is a cell line that provides an ideal cell system to propagate small airway epithelial cells in serum-free conditions when grown in Airway Epithelial Cell Basal Media supplemented with Bronchial Epithelial Cell Growth Kit (ATCC PCS-300-040) components.
Catalog No. 50-238-4580 Supplier ATCC Supplier No. PCS301010
Primary Small Airway Epithelial Cells; Normal, Human (HSAEC) is a cell line that provides an ideal cell system to propagate small airway epithelial cells in serum-free conditions when grown in Airway Epithelial Cell Basal Media supplemented with Bronchial Epithelial Cell Growth Kit (ATCC PCS-300-040) components.