Learn More
Hach Company Nitrogen Primary Standard, set of 3
Supplier: Hach Company 2277800
Use this set for checking accuracy of crude protein tests and digestion recovery efficiency in Total Nitrogen and TKN methods. Set of three 25-g standards: Ammonium p-Toluenesulfonic Acid (Ammonium PTSA), Glycine PTSA and Nicotinic Acid PTSA.