Learn More
ATCC Primary Subcutaneous Pre-adipocytes Normal Human
Catalog No. 502382390
Encompass
Normal Human Pre-Adipocytes were isolated from the adipose tissue of a donor and are derived from de-differentiated mature adipocytes by ceiling culture and providing an ideal culture model for the study of diabetes, obesity, metabolism, insulin sensitivity, and adipose biology.
Catalog No. 50-238-2390 Supplier ATCC Supplier No. PCS210010
Normal Human Pre-Adipocytes were isolated from the adipose tissue of a donor and are derived from de-differentiated mature adipocytes by ceiling culture and providing an ideal culture model for the study of diabetes, obesity, metabolism, insulin sensitivity, and adipose biology.