Learn More
ATCC Primary Umbilical Vein Endothelial Cells; Normal, Human
Catalog No. 502384144
Encompass
Normal Primary Human Umbilical Vein Endothelial Cells (HUVEC), when grown in Vascular Cell Basal Media supplemented with Endothelial Cell Growth Kit components, provide an ideal cell system to propagate HUVEC in low serum conditions with or without the addition of human recombinant VEGF.
Catalog No. 50-238-4144 Supplier ATCC Supplier No. PCS100010
Normal Primary Human Umbilical Vein Endothelial Cells (HUVEC), when grown in Vascular Cell Basal Media supplemented with Endothelial Cell Growth Kit components, provide an ideal cell system to propagate HUVEC in low serum conditions with or without the addition of human recombinant VEGF.