Learn More
Cayman Chemical Primary Vascular Eicosanoid MaxSpec LC-MS Mixture, 1 ea
SDP
Catalog No. 502984550
Encompass_Preferred
Contains a collection of vasoactive eicosanoids, including the characteristic metabolites of both PGI2 and TXA2 and several additional metabolites produced by platelets and the CYP450 pathway of arachidonic acid metabolism; designed for direct use in LC-MS applications
Catalog No. 50-298-4550 Supplier Cayman Chemical Supplier No. 196671 EA
Contains a collection of vasoactive eicosanoids, including the characteristic metabolites of both PGI2 and TXA2 and several additional metabolites produced by platelets and the CYP450 pathway of arachidonic acid metabolism; designed for direct use in LC-MS applications