Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Monkey TNF-alpha ELISA Kit (Colorimetric)
SDP

Catalog No. NBP194190
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP194190 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP194190 Supplier Novus Biologicals™ Supplier No. NBP194190

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.

The Monkey TNF-alpha ELISA Kit (Colorimetric) measures monkey TNF-alpha in cell culture supernatant, serum (baboon, cynomolgus).

Specifications

Assay Range 28.1 - 1,800 U/mL
Assay Sensitivity 6.0 U/mL
Product Type ELISA Kit
Sample Type Baboon Serum, Cell Culture Supernatant, Cynomolgus Serum
Target Species Primate
For Use With (Application) ELISA
Gene ID (Entrez) 7124
Gene Symbol TNF
Kit Contents Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to monkey TNF-alpha, Biotin-Conjugate anti-monkey TNF-alpha polyclonal antibody, Streptavidin-HRP, Monkey TNF-alpha Standard lyophilized, 3600U/mL upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
Quantity 96 Tests
Research Discipline Adaptive Immunity, Asthma, Autophagy, Biologically Active Proteins, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necr
Target TNF-alpha
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.