Learn More
Leica Biosystems Prime bottle assembly - replacement part for Leica Biosystems instruments
Catalog No. NC3844357
Encompass
The Prime bottle assembly is a replacement assembly used to prime fluid and reagent lines in automated laboratory instruments. Designed as a spare or service part, it helps restore proper fluid flow and reduce air entrainment during reagent loading.
  • Replacement bottle assembly for priming reagent lines.
  • Restores proper fluid delivery and system priming.
  • Reduces air entrainment during reagent loading.
  • Simple installation for service technicians.
  • Sold as a single assembly for use as a spare part.
Catalog No. NC3844357 Supplier Leica Biosystems Supplier No. 14051453931
The Prime bottle assembly is a replacement assembly used to prime fluid and reagent lines in automated laboratory instruments. Designed as a spare or service part, it helps restore proper fluid flow and reduce air entrainment during reagent loading.
  • Replacement bottle assembly for priming reagent lines.
  • Restores proper fluid delivery and system priming.
  • Reduces air entrainment during reagent loading.
  • Simple installation for service technicians.
  • Sold as a single assembly for use as a spare part.