Learn More
Nova Biomedical Corporation Nova Biomedical Prime Calibrator Cartridge, Individual Maintenance-Free, Rapid Replacement, 1 unit
Catalog No. NC2090064
Encompass
The Nova Biomedical Prime Calibrator Cartridge is essential for precise measurements in critical care blood gas testing. Its rapid replacement system minimizes downtime, while RFID technology ensures accurate tracking of usage and history. This maintenance-free, individual cartridge design reduces laboratory waste compared to combined systems, offering a cost-effective solution. Ideal for various analytes, it ensures reliable results with high efficiency.
This item is not returnable. View return policy
Catalog No. NC2090064 Supplier Nova Biomedical Corporation Supplier No. 53365
 Restricted Item
Add to Cart
Add to Cart
The Nova Biomedical Prime Calibrator Cartridge is essential for precise measurements in critical care blood gas testing. Its rapid replacement system minimizes downtime, while RFID technology ensures accurate tracking of usage and history. This maintenance-free, individual cartridge design reduces laboratory waste compared to combined systems, offering a cost-effective solution. Ideal for various analytes, it ensures reliable results with high efficiency.