Learn More
Nova Biomedical Corporation Nova Biomedical PRIME PLUS AUTO QC W/BUN CRERX, Automated Quality Control, Adheres to Regulatory Standards, 22.65 units
Catalog No. NC2064446
Encompass
The PRIME PLUS AUTO QC W/BUN CRERX is an automated liquid quality control cartridge designed for use with critical care analyzers. It ensures reliable analytical performance by verifying important assays like blood urea nitrogen (BUN) and creatinine, promoting compliance with stringent regulatory standards. Its maintenance-free design enhances uptime, providing real-time assurance for laboratories seeking efficiency and accuracy during quality control processes.
This item is not returnable. View return policy
Catalog No. NC2064446 Supplier Nova Biomedical Corporation Supplier No. 57841
The PRIME PLUS AUTO QC W/BUN CRERX is an automated liquid quality control cartridge designed for use with critical care analyzers. It ensures reliable analytical performance by verifying important assays like blood urea nitrogen (BUN) and creatinine, promoting compliance with stringent regulatory standards. Its maintenance-free design enhances uptime, providing real-time assurance for laboratories seeking efficiency and accuracy during quality control processes.