Learn More
Nova Biomedical Corporation Nova Biomedical Stat Profile Prime Plus Calibrator Cartridge, Maintenance-Free, 100 Sample Capacity, RFID Technology, Each
Catalog No. NC2182233
Encompass
The Nova Biomedical Stat Profile Prime Plus Calibrator Cartridge is a maintenance-free component designed for the Stat Profile Prime Plus blood gas analyzer. It ensures optimal performance with a capacity for 100 samples, enabling accurate measurements of Creatinine and BUN. Its quick replacement process and integrated RFID technology enhance analyzer uptime and inventory management, providing a significant advantage over traditional systems.
This item is not returnable. View return policy
Catalog No. NC2182233 Supplier Nova Biomedical Corporation Supplier No. 57831
The Nova Biomedical Stat Profile Prime Plus Calibrator Cartridge is a maintenance-free component designed for the Stat Profile Prime Plus blood gas analyzer. It ensures optimal performance with a capacity for 100 samples, enabling accurate measurements of Creatinine and BUN. Its quick replacement process and integrated RFID technology enhance analyzer uptime and inventory management, providing a significant advantage over traditional systems.