Learn More
Waters Corp Prime/Purge Valve 700000148
Supplier:  Waters Corp 700000148
Encompass_Preferred
The prime/purge valve vents the flow to atmosphere and permits efficient priming of the WatersReagent Manager pump.
Catalog No. 50-792-315