Learn More
Waters Corp WATERS CORPORATION Prime/Vent Valve, Replacement, for HPLC Alliance Separation Modules
Catalog No. 50793822 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The WAT270965 is a Replacement Prime/Vent Valve Assembly. This valve is designed to receive flow from the system pressure transducer outlet. From the vent valve, the flow proceeds into an in-line filter. It is used in Alliance Separation Modules.
  • Replacement prime/vent valve assembly.
  • Receives flow from the system pressure transducer outlet.
  • Flow proceeds into an in-line filter.
  • Used in Alliance Separation Modules.
Catalog No. 50-793-822 Supplier Waters Corp Supplier No. WAT270965
The WAT270965 is a Replacement Prime/Vent Valve Assembly. This valve is designed to receive flow from the system pressure transducer outlet. From the vent valve, the flow proceeds into an in-line filter. It is used in Alliance Separation Modules.
  • Replacement prime/vent valve assembly.
  • Receives flow from the system pressure transducer outlet.
  • Flow proceeds into an in-line filter.
  • Used in Alliance Separation Modules.