Learn More
Waters Corp Waters Prime/Vent Valve Rebuild Kit, HPLC
Catalog No. 50793815 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit contains a replacement seal and valve tip to rebuild an HPLC Prime/Purge Valve.
  • Type: Rebuild kit
  • System type: HPLC
  • Replacement schedule: As required
  • Units per package: 1
Catalog No. 50-793-815 Supplier Waters Corp Supplier No. WAT270936
This kit contains a replacement seal and valve tip to rebuild an HPLC Prime/Purge Valve.
  • Type: Rebuild kit
  • System type: HPLC
  • Replacement schedule: As required
  • Units per package: 1