Learn More
FUJIFILM Biosciences PRIME-XV Dendritic Cell Maturation CDM, 500 mL
Catalog No. NC1745128
Encompass_Preferred
PRIME-XV Dendritic Cell Maturation CDM is a chemically-defined, animal component-free medium developed for the in vitro differentiation of monocytes into immature dendritic cells (iDCs) and subsequent maturation to produce mature dendritic cells (mDCs). Free of the undefined components found in animal- or human-derived raw materials that affect functional capacity PRIME-XV Dendritic Cell Maturation CDM supports effective maturation of mDCs with the functional capacity to induce T cell response.
Catalog No. NC1745128 Supplier FUJIFILM Biosciences Supplier No. 91146500ML
PRIME-XV Dendritic Cell Maturation CDM is a chemically-defined, animal component-free medium developed for the in vitro differentiation of monocytes into immature dendritic cells (iDCs) and subsequent maturation to produce mature dendritic cells (mDCs). Free of the undefined components found in animal- or human-derived raw materials that affect functional capacity PRIME-XV Dendritic Cell Maturation CDM supports effective maturation of mDCs with the functional capacity to induce T cell response.