Learn More
FUJIFILM Biosciences PRIME-XV T Cell CDM 1l
Catalog No. 502892206
Encompass_Preferred
PRIME-XV T Cell CDM is the first commercially-available, chemically defined, animal component-free medium for the expansion and transduction of human T cells. The formulation is optimized to deliver consistently vigorous growth while maintaining T cell functionality and potency.Supports T cell expansion in static and dynamic automation systems while maintaining functionalityEliminates the adverse effects undefined components cause on T cell phenotypesSupports polarization to targeted T cell types, such as Th1, Th2, cytotoxic T cells, to further possible therapy applicationsPerforms transduction of T cells without requiring spinoculation or transduction enhancers
Catalog No. 50-289-2206 Supplier FUJIFILM Biosciences Supplier No. 911541L
 Restricted Item
Add to Cart
Add to Cart
PRIME-XV T Cell CDM is the first commercially-available, chemically defined, animal component-free medium for the expansion and transduction of human T cells. The formulation is optimized to deliver consistently vigorous growth while maintaining T cell functionality and potency.Supports T cell expansion in static and dynamic automation systems while maintaining functionalityEliminates the adverse effects undefined components cause on T cell phenotypesSupports polarization to targeted T cell types, such as Th1, Th2, cytotoxic T cells, to further possible therapy applicationsPerforms transduction of T cells without requiring spinoculation or transduction enhancers