Learn More
FUJIFILM Biosciences PRIME-XV T CELL CDM
Catalog No. 5000812419
Encompass_Preferred
PRIME-XV T Cell CDM is the first commercially-available chemically defined animal component-free medium for the expansion and transduction of human T cells The formulation is optimized to deliver consistently vigorous growth while maintaining T cell functionality and potency Provides Optimal PerformanceChemically defined animal component-free T cell culture mediumSupports T cell expansion in static and dynamic automation systems while maintaining functionalityEliminates the adverse effects undefined components cause on T cell phenotypesSupports polarization to targeted T cell types such as Th1 Th2 cytotoxic T cells to further possible therapy applicationsPerforms transduction of T cells without requiring spinoculation or transduction enhancers
Catalog No. 50-008-12419 Supplier FUJIFILM Biosciences Supplier No. 911545L
PRIME-XV T Cell CDM is the first commercially-available chemically defined animal component-free medium for the expansion and transduction of human T cells The formulation is optimized to deliver consistently vigorous growth while maintaining T cell functionality and potency Provides Optimal PerformanceChemically defined animal component-free T cell culture mediumSupports T cell expansion in static and dynamic automation systems while maintaining functionalityEliminates the adverse effects undefined components cause on T cell phenotypesSupports polarization to targeted T cell types such as Th1 Th2 cytotoxic T cells to further possible therapy applicationsPerforms transduction of T cells without requiring spinoculation or transduction enhancers