Learn More
FUJIFILM Biosciences PRIME-XV T CELL EXPANSION XSFM
Catalog No. 5000812433
Encompass_Preferred
PRIME-XV T Cell XSFM is a xeno-free and serum-free culture medium that supports the expansion and reactivation of human T cells as well as the transition from preclinical to clinical applications for immunotherapy therapeutics Provides Optimal PerformanceXeno-free serum-free medium for T cell cultureOptimized for activation and expansion of human T lymphocytesMaintains potency of major T cells subsets such as CD4 and CD8 Supports expansion of re-activated T cellsSerum-free and xeno-free formulation reduces risk of contamination and provides lot-to-lot consistencyContains gentamicin for reduced contamination of culture componentsDesigned and manufactured to facilitate transfer from research to clinic Supported by robust raw material controls and supply chain management
Catalog No. 50-008-12433 Supplier FUJIFILM Biosciences Supplier No. 911415L
PRIME-XV T Cell XSFM is a xeno-free and serum-free culture medium that supports the expansion and reactivation of human T cells as well as the transition from preclinical to clinical applications for immunotherapy therapeutics Provides Optimal PerformanceXeno-free serum-free medium for T cell cultureOptimized for activation and expansion of human T lymphocytesMaintains potency of major T cells subsets such as CD4 and CD8 Supports expansion of re-activated T cellsSerum-free and xeno-free formulation reduces risk of contamination and provides lot-to-lot consistencyContains gentamicin for reduced contamination of culture componentsDesigned and manufactured to facilitate transfer from research to clinic Supported by robust raw material controls and supply chain management