Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PrimeFlow™ RNA Assay Kit

Catalog No. 8818005204
Encompass_Preferred
Change view
Click to view available options
No. of Reactions:
40 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
88-180-05204 40 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 88-180-05204 Supplier Invitrogen™ Supplier No. 8818005204
Only null left
Add to Cart
Add to Cart

An in situ hybridization assay that combines the power of branched-DNA technology with the single-cell resolution of flow cytometry, enabling the detection of four RNA targets in combination with immunophenotyping for cell surface and intracellular proteins using fluorochrome-conjugated antibodies.

This PrimeFlow™ RNA Assay is an in situ hybridization assay that combines the power of branched-DNA technology with the single-cell resolution of flow cytometry, enabling the simultaneous detection of up to four RNA targets in combination with immunophenotyping for cell surface and intracellular proteins using fluorochrome-conjugated antibodies. Branched-DNA technology uses pairs of target-specific probes to amplify the detection of a RNA transcript, achieving between 8,000- to 16,000-fold signal amplification from a desired RNA transcript. PrimeFlow RNA Assay can detect mRNA, long non-coding RNA, viral RNA, and telomere DNA. The detection of microRNA is also possible and is optimal when using the PrimeFlow™ microRNA Pretreatment Buffer (cat. 88-18006) and protocol. PrimeFlow RNA Assay is compatible with human and mouse cells. Other mammalian and non-mammalian cells may be compatible

This PrimeFlow RNA Assay contains all the reagents needed to conduct the assay. Positive control probes and target probes for genes of interest are sold separately.

Reported Application
RNA Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Detection Method Primer-probe Detection
Format Kit
No. of Reactions 40 Tests
Product Type RNA In Situ Hybridization Assay Kit
Quantity 40 Tests

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.