Learn More
Agilent Technologies PRIMEIT2 RANDPRIMERLABELKIT
Catalog No. 50125542
Encompass
Prime-It II Random Primer Labeling Kit. Produces high-specific-activity probes in 2 minutes. Contains: Exo-Klenow polymerase, Random 9-mer primers, Control template, Reaction buffers for [32P]dATP and [32P]dCTP labeling, Stop mix
Catalog No. 50-125-542 Supplier Agilent Technologies Supplier No. 300385
Prime-It II Random Primer Labeling Kit. Produces high-specific-activity probes in 2 minutes. Contains: Exo-Klenow polymerase, Random 9-mer primers, Control template, Reaction buffers for [32P]dATP and [32P]dCTP labeling, Stop mix